Search
Search

BIRC8
FAM90A27PAGACATAAAAACTCTTTGCTTGAAA[C/T]CAATAACCGCGCTGCACATGGGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BIRC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| BIRC8 - baculoviral IAP repeat containing 8 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_033341.4 | Intron | NP_203127.3 | ||||
| FAM90A27P - family with sequence similarity 90, member A27, pseudogene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||