Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCAGGCTCCACCAGGACTCGCC[A/G]CCCTGAGATCTCCCAGGCTGGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102670 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MADCAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MADCAM1 - mucosal vascular addressin cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130760.2 | 885 | Missense Mutation | CAC,CGC | H,R 292 | NP_570116.2 | |
NM_130762.2 | 885 | Intron | NP_570118.1 |
TPGS1 - tubulin polyglutamylase complex subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |