Search
Search

ICAM1
ICAM4
ICAM5TAGTCCGGGCTTTTTGCCATGGGGT[C/T]TCTGTTCCCTCTGTCGCTGCTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ICAM1 - intercellular adhesion molecule 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000201.2 | 54 | Intron | NP_000192.2 | |||
| ICAM4 - intercellular adhesion molecule 4 (Landsteiner-Wiener blood group) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001039132.2 | 54 | Missense Mutation | TCT,TTT | S,F 3 | NP_001034221.1 | |
| NM_001544.4 | 54 | Missense Mutation | TCT,TTT | S,F 3 | NP_001535.1 | |
| NM_022377.3 | 54 | Missense Mutation | TCT,TTT | S,F 3 | NP_071772.1 | |
| ICAM5 - intercellular adhesion molecule 5 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||