Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AAAGCCTTCAGAGCTGTGTCAATCC[C/T]TTGAATGCATGGTAGGACTCACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF440 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF440 - zinc finger protein 440 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152357.2 | 1622 | Missense Mutation | CTT,TTT | L,F 530 | NP_689570.2 | |
XM_005259731.3 | 1622 | Missense Mutation | CTT,TTT | L,F 533 | XP_005259788.1 | |
XM_017026253.1 | 1622 | Missense Mutation | CTT,TTT | L,F 532 | XP_016881742.1 | |
XM_017026254.1 | 1622 | Missense Mutation | CTT,TTT | L,F 408 | XP_016881743.1 |