Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GCCACCTGCACCAATACAGTGGACA[C/G]TTACTATTGCGCTTGCAAACAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600493 | ||||||||||||||||||||
Literature Links: |
ADGRE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRE1 - adhesion G protein-coupled receptor E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256252.1 | 218 | Missense Mutation | ACT,AGT | T,S 53 | NP_001243181.1 | |
NM_001256253.1 | 218 | Missense Mutation | ACT,AGT | T,S 53 | NP_001243182.1 | |
NM_001256254.1 | 218 | Missense Mutation | ACT,AGT | T,S 53 | NP_001243183.1 | |
NM_001256255.1 | 218 | Missense Mutation | ACT,AGT | T,S 53 | NP_001243184.1 | |
NM_001974.4 | 218 | Missense Mutation | ACT,AGT | T,S 53 | NP_001965.3 | |
XM_011527794.1 | 218 | Missense Mutation | ACT,AGT | T,S 53 | XP_011526096.1 |