Search Thermo Fisher Scientific
GTATCCAGGGAGAAGTGGACGTGCT[C/G]GGTGAGACTTCACAGCCCCTGATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120216 MIM: 608575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL5A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL5A3 - collagen type V alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RDH8 - retinol dehydrogenase 8 (all-trans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015725.2 | 510 | Silent Mutation | CTC,CTG | L,L 107 | NP_056540.2 |