Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GGCGGGGCTGGCGGCGGCCCACAGC[G/T]TGCGCCTGGCGCGCGGGGACCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609762 MIM: 610396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLOC1S3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLOC1S3 - biogenesis of lysosomal organelles complex 1 subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_212550.4 | 574 | Missense Mutation | GTG,TTG | V,L 160 | NP_997715.1 |
TRAPPC6A - trafficking protein particle complex 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270891.1 | 574 | Intron | NP_001257820.1 | |||
NM_001270892.1 | 574 | Intron | NP_001257821.1 | |||
NM_001270893.1 | 574 | Intron | NP_001257822.1 | |||
NM_024108.2 | 574 | Intron | NP_077013.1 |