Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGAACAGAGCAGAAACGGTGATAAT[T/G]ATCTGTATGAAAAAGTAAGGATACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 191030 | ||||||||||||||||||||
Literature Links: |
TPM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPM3 - tropomyosin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001043351.1 | Intron | NP_001036816.1 | ||||
NM_001043352.1 | Intron | NP_001036817.1 | ||||
NM_001043353.1 | Intron | NP_001036818.1 | ||||
NM_001278188.1 | Intron | NP_001265117.1 | ||||
NM_001278189.1 | Intron | NP_001265118.1 | ||||
NM_001278190.1 | Intron | NP_001265119.1 | ||||
NM_001278191.1 | Intron | NP_001265120.1 | ||||
NM_153649.3 | Intron | NP_705935.1 |