Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGTATCTGCCGGCCCTTGGTC[C/T]GGTGAGAAGGCAGAGATGAACATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612992 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBPF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NBPF3 - neuroblastoma breakpoint family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256416.2 | Intron | NP_001243345.1 | ||||
NM_001256417.2 | Intron | NP_001243346.1 | ||||
NM_032264.4 | Intron | NP_115640.1 | ||||
XM_006710957.1 | Intron | XP_006711020.1 | ||||
XM_006710958.1 | Intron | XP_006711021.1 | ||||
XM_006710959.2 | Intron | XP_006711022.1 | ||||
XM_011542276.1 | Intron | XP_011540578.1 | ||||
XM_011542277.1 | Intron | XP_011540579.1 | ||||
XM_011542278.1 | Intron | XP_011540580.1 | ||||
XM_011542281.1 | Intron | XP_011540583.1 | ||||
XM_017002498.1 | Intron | XP_016857987.1 | ||||
XM_017002499.1 | Intron | XP_016857988.1 | ||||
XM_017002500.1 | Intron | XP_016857989.1 | ||||
XM_017002501.1 | Intron | XP_016857990.1 | ||||
XM_017002502.1 | Intron | XP_016857991.1 |