Search
Search

CPSF3L
CPTP
DVL1
TAS1R3CCCACGAGCTGAGCAACCGGCTGGT[A/G]GACCTGGAGCGCCGCTCCCACCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF3L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CPSF3L - cleavage and polyadenylation specific factor 3-like | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| CPTP - ceramide-1-phosphate transfer protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001029885.1 | 691 | Silent Mutation | GTA,GTG | V,V 92 | NP_001025056.1 | |
| XM_005244801.3 | 691 | Silent Mutation | GTA,GTG | V,V 92 | XP_005244858.1 | |
| XM_005244802.1 | 691 | Silent Mutation | GTA,GTG | V,V 29 | XP_005244859.1 | |
| XM_011542200.2 | 691 | Silent Mutation | GTA,GTG | V,V 92 | XP_011540502.1 | |
| DVL1 - dishevelled segment polarity protein 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| TAS1R3 - taste 1 receptor member 3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||