Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CCCAAGTGCCAGGAGAGCTTTCGGC[A/G]GCGCTCAGACCTCACCACGCACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609017 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERMAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERMAP - erythroblast membrane associated protein (Scianna blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC339539 - uncharacterized LOC339539 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF691 - zinc finger protein 691 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242739.1 | 4529 | Missense Mutation | CAG,CGG | Q,R 182 | NP_001229668.1 | |
NM_015911.3 | 4529 | Missense Mutation | CAG,CGG | Q,R 151 | NP_056995.1 | |
XM_006710671.3 | 4529 | Missense Mutation | CAG,CGG | Q,R 178 | XP_006710734.1 | |
XM_017001400.1 | 4529 | Missense Mutation | CAG,CGG | Q,R 182 | XP_016856889.1 | |
XM_017001401.1 | 4529 | Missense Mutation | CAG,CGG | Q,R 182 | XP_016856890.1 | |
XM_017001402.1 | 4529 | Missense Mutation | CAG,CGG | Q,R 173 | XP_016856891.1 | |
XM_017001403.1 | 4529 | Missense Mutation | CAG,CGG | Q,R 173 | XP_016856892.1 |