Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TCAGAACGGTGAAGATTGATGTGTA[C/T]GACTGGGACCGGGATGGAAGGTAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CPNE9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPNE9 - copine family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308388.1 | 1019 | Silent Mutation | TAC,TAT | Y,Y 224 | NP_001295317.1 | |
NM_153635.2 | 1019 | Silent Mutation | TAC,TAT | Y,Y 224 | NP_705899.2 | |
XM_011533386.2 | 1019 | Silent Mutation | TAC,TAT | Y,Y 333 | XP_011531688.1 | |
XM_011533388.2 | 1019 | Silent Mutation | TAC,TAT | Y,Y 333 | XP_011531690.1 | |
XM_011533389.2 | 1019 | Silent Mutation | TAC,TAT | Y,Y 333 | XP_011531691.1 | |
XM_017005747.1 | 1019 | UTR 5 | XP_016861236.1 |