Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCGGATGTTGAGCAGGTCAAT[C/T]TCACTGCGGGATACCATGATCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114070 | ||||||||||||||||||||
Literature Links: |
ANXA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANXA6 - annexin A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001155.4 | 2103 | Silent Mutation | GAA,GAG | E,E 631 | NP_001146.2 | |
NM_001193544.1 | 2103 | Silent Mutation | GAA,GAG | E,E 599 | NP_001180473.1 | |
XM_005268432.4 | 2103 | Silent Mutation | GAA,GAG | E,E 625 | XP_005268489.1 |