Search
Search

DEFB113
DEFB114GTTAATGATTGGCTTTTGGTATTCC[C/T]ATACCGCACAGCAGGGGTTAACATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
DEFB113 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| DEFB113 - defensin beta 113 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001037729.1 | 194 | Nonsense Mutation | TAG,TGG | *,W 65 | NP_001032818.1 | |
| DEFB114 - defensin beta 114 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||