Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
CCAGGCTCACCCCCTTGTCTGCTGC[A/G]TAGTTAGAGTTGCTCACAAAGGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611499 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUSB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUSB - glucuronidase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000181.3 | 1336 | Silent Mutation | TAC,TAT | Y,Y 487 | NP_000172.2 | |
NM_001284290.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 341 | NP_001271219.1 | |
NM_001293104.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 297 | NP_001280033.1 | |
NM_001293105.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 268 | NP_001280034.1 | |
XM_005250297.3 | 1336 | Silent Mutation | TAC,TAT | Y,Y 436 | XP_005250354.1 | |
XM_011516113.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 320 | XP_011514415.1 | |
XM_011516114.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 263 | XP_011514416.1 | |
XM_017012091.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 269 | XP_016867580.1 | |
XM_017012092.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 246 | XP_016867581.1 | |
XM_017012093.1 | 1336 | Silent Mutation | TAC,TAT | Y,Y 212 | XP_016867582.1 |