Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GTCACTCTCCTCGAAGATCTGGAGC[A/G]GGAACTGGATGAGCCAGGACACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194510 MIM: 601261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF3 - zinc finger protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN21 - zinc finger and SCAN domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145914.2 | 518 | Missense Mutation | CAG,CGG | Q,R 125 | NP_666019.1 | |
XM_005250568.4 | 518 | Missense Mutation | CAG,CGG | Q,R 125 | XP_005250625.1 | |
XM_006716113.3 | 518 | Missense Mutation | CAG,CGG | Q,R 125 | XP_006716176.1 | |
XM_017012584.1 | 518 | Missense Mutation | CAG,CGG | Q,R 125 | XP_016868073.1 | |
XM_017012585.1 | 518 | Missense Mutation | CAG,CGG | Q,R 125 | XP_016868074.1 |