Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GATAGAAGACTTACAGAGAGAACTT[A/G]AGATACCAGAGCAGCAGGTAAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300627 MIM: 314997 | ||||||||||||||||||||
Literature Links: |
ZNF449 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF449 - zinc finger protein 449 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152695.5 | 502 | Missense Mutation | AAG,GAG | K,E 113 | NP_689908.3 | |
XM_011531312.2 | 502 | Missense Mutation | AAG,GAG | K,E 113 | XP_011529614.1 | |
XM_011531313.2 | 502 | Intron | XP_011529615.1 | |||
XM_011531314.2 | 502 | UTR 5 | XP_011529616.1 | |||
XM_017029350.1 | 502 | UTR 5 | XP_016884839.1 | |||
XM_017029351.1 | 502 | Intron | XP_016884840.1 | |||
XM_017029352.1 | 502 | Intron | XP_016884841.1 |
ZNF75D - zinc finger protein 75D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |