Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGGGGAGCAGGCCCCCACGCA[A/G]AATGAAGAAGAATCCCGCCATTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEX4 - brain expressed X-linked 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080425.3 | 242 | Silent Mutation | CAA,CAG | Q,Q 30 | NP_001073894.1 | |
NM_001127688.2 | 242 | Silent Mutation | CAA,CAG | Q,Q 30 | NP_001121160.1 |