Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2C9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
Caucasian
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
African American
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
African American
|
JPT (Japanese) - Not Available | ||||||
EUR
|
Japanese
|
|||||||
AMR
|
Japanese
|
|||||||
Chinese
|
||||||||
Chinese
|
| CYP2C9 - cytochrome P450 family 2 subfamily C member 9 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000771.3 | 474 | Missense Mutation | CAC,CGC | H,R 150 | NP_000762.2 | |
| XM_017015758.1 | 474 | Missense Mutation | CAC,CGC | H,R 150 | XP_016871247.1 | |
Set Membership: |
|