Search
Search

CNOT7
ZDHHC2TAGTCTGATACTGTCAGTCTATTCT[T/C]TTACCAGTTATGGTGACACTGCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
CNOT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| CNOT7 - CCR4-NOT transcription complex subunit 7 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001322087.1 | 4291 | Intron | NP_001309016.1 | |||
| NM_001322088.1 | 4291 | Intron | NP_001309017.1 | |||
| NM_001322089.1 | 4291 | Intron | NP_001309018.1 | |||
| NM_001322090.1 | 4291 | Intron | NP_001309019.1 | |||
| NM_001322091.1 | 4291 | Intron | NP_001309020.1 | |||
| NM_001322092.1 | 4291 | Intron | NP_001309021.1 | |||
| NM_001322093.1 | 4291 | Intron | NP_001309022.1 | |||
| NM_001322094.1 | 4291 | Intron | NP_001309023.1 | |||
| NM_001322095.1 | 4291 | Intron | NP_001309024.1 | |||
| NM_001322096.1 | 4291 | Intron | NP_001309025.1 | |||
| NM_001322097.1 | 4291 | Intron | NP_001309026.1 | |||
| NM_001322098.1 | 4291 | Intron | NP_001309027.1 | |||
| NM_001322099.1 | 4291 | Intron | NP_001309028.1 | |||
| NM_001322100.1 | 4291 | Intron | NP_001309029.1 | |||
| NM_013354.6 | 4291 | Intron | NP_037486.2 | |||
| NM_054026.3 | 4291 | Intron | NP_473367.2 | |||
| XM_005273481.2 | 4291 | UTR 3 | XP_005273538.1 | |||
| ZDHHC2 - zinc finger DHHC-type containing 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_016353.4 | 4291 | Intron | NP_057437.1 | |||