Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_308362630_10
          See other BNC2 GT Assays ›
          SNP ID:
          rs377056909
          Gene
          BNC2
          Gene Name
          basonuclin 2
          Set Membership:
          -
          Chromosome Location:
          Chr.9: 16437210 - 16437210 on Build GRCh38
          Polymorphism:
          T/C, Transition substitution
          Context Sequence [VIC/FAM]:

          CTAGCAGTGGTGCTGAGACAGGGTT[T/C]ATGTACTGGAATGGAAGCAGAAATG

          Assay ID C_308362630_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 608669

          Literature Links:

          BNC2 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          BNC2 - basonuclin 2
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001317939.1 5813 Missense Mutation ATA,ATG I,M 286 NP_001304868.1
          NM_001317940.1 5813 Missense Mutation ATA,ATG I,M 233 NP_001304869.1
          NM_017637.5 5813 Missense Mutation ATA,ATG I,M 328 NP_060107.3
          XM_011517924.1 5813 Missense Mutation ATA,ATG I,M 286 XP_011516226.1
          XM_011517934.2 5813 Missense Mutation ATA,ATG I,M 154 XP_011516236.1
          XM_017014816.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870305.1
          XM_017014817.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870306.1
          XM_017014818.1 5813 Missense Mutation ATA,ATG I,M 342 XP_016870307.1
          XM_017014819.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870308.1
          XM_017014820.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870309.1
          XM_017014821.1 5813 Missense Mutation ATA,ATG I,M 342 XP_016870310.1
          XM_017014822.1 5813 Missense Mutation ATA,ATG I,M 219 XP_016870311.1
          XM_017014823.1 5813 Missense Mutation ATA,ATG I,M 219 XP_016870312.1
          XM_017014824.1 5813 Missense Mutation ATA,ATG I,M 154 XP_016870313.1
          XM_017014825.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870314.1
          XM_017014826.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870315.1
          XM_017014827.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870316.1
          XM_017014828.1 5813 Missense Mutation ATA,ATG I,M 370 XP_016870317.1
          XM_017014829.1 5813 Missense Mutation ATA,ATG I,M 93 XP_016870318.1

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          endochondral bone growth
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          tongue development
          palate development
          mesenchyme development
          DNA binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0