Search
Search

KAT5
RNASEH2CGGCTCCTGGGGACAGATAAAGGTCC[T/C]CAGGGAACCTGACCTGTGCTCTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
KAT5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| KAT5 - lysine acetyltransferase 5 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001206833.1 | 1615 | Intron | NP_001193762.1 | |||
| NM_006388.3 | 1615 | Intron | NP_006379.2 | |||
| NM_182709.2 | 1615 | Intron | NP_874368.1 | |||
| NM_182710.2 | 1615 | Intron | NP_874369.1 | |||
| XM_006718421.2 | 1615 | Intron | XP_006718484.1 | |||
| RNASEH2C - ribonuclease H2 subunit C | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_032193.3 | 1615 | UTR 3 | NP_115569.2 | |||