Search
Search

TMEM45BCCCAGGAGGACAGACAGCCAGTTCA[A/G]TTTTAAAGAGAATATTCTCAAAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM45B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| TMEM45B - transmembrane protein 45B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_138788.3 | Intron | NP_620143.1 | ||||
| XM_005271400.4 | Intron | XP_005271457.1 | ||||
| XM_011542586.2 | Intron | XP_011540888.1 | ||||
| XM_011542588.2 | Intron | XP_011540890.1 | ||||
| XM_017017188.1 | Intron | XP_016872677.1 | ||||
| XM_017017189.1 | Intron | XP_016872678.1 | ||||