Search
Search

HIKESHIGGAGAGAGCAATATTTAAAGCTAAA[A/G]ATTGTTTAGAATTGATGATAAATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
HIKESHI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| HIKESHI - Hikeshi, heat shock protein nuclear import factor | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001322404.1 | Intron | NP_001309333.1 | ||||
| NM_001322407.1 | Intron | NP_001309336.1 | ||||
| NM_001322409.1 | Intron | NP_001309338.1 | ||||
| NM_016401.3 | Intron | NP_057485.2 | ||||
| XM_017017914.1 | Intron | XP_016873403.1 | ||||
| XM_017017915.1 | Intron | XP_016873404.1 | ||||