Search
Search

DHRS2ACACATGACCTCCTGTCTGCAGGAT[G/T]CAAGTTCCCTCTACAGAGTCAGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
DHRS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| DHRS2 - dehydrogenase/reductase 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001318835.1 | Intron | NP_001305764.1 | ||||
| NM_005794.3 | Intron | NP_005785.1 | ||||
| NM_182908.4 | Intron | NP_878912.1 | ||||
| XM_005267249.1 | Intron | XP_005267306.1 | ||||
| XM_006720001.3 | Intron | XP_006720064.1 | ||||
| XM_011536338.1 | Intron | XP_011534640.1 | ||||
| XM_011536339.2 | Intron | XP_011534641.1 | ||||