Search
Search

CYB561
TANC2AGATCTGTGCTCTGCGACCAGGACC[G/A]AACACCCGAAGTCCTACCCAAAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
CYB561 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| CYB561 - cytochrome b561 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001017916.1 | 1199 | UTR 3 | NP_001017916.1 | |||
| NM_001017917.1 | 1199 | UTR 3 | NP_001017917.1 | |||
| NM_001915.3 | 1199 | UTR 3 | NP_001906.3 | |||
| XM_005257091.1 | 1199 | UTR 3 | XP_005257148.1 | |||
| XM_017024260.1 | 1199 | UTR 3 | XP_016879749.1 | |||
| XM_017024261.1 | 1199 | UTR 3 | XP_016879750.1 | |||
| XM_017024262.1 | 1199 | UTR 3 | XP_016879751.1 | |||
| TANC2 - tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||