Search
Search

EML2GCGCCCAGCACCGCCTGTGTGATAC[G/A]GTTCCCACCTGCAGGGTGGCCAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EML2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| EML2 - echinoderm microtubule associated protein like 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001193268.1 | 1607 | Missense Mutation | CGT,TGT | R,C 485 | NP_001180197.1 | |
| NM_001193269.1 | 1607 | Missense Mutation | CGT,TGT | R,C 431 | NP_001180198.1 | |
| NM_012155.2 | 1607 | Missense Mutation | CGT,TGT | R,C 284 | NP_036287.1 | |
| XM_006723125.2 | 1607 | Missense Mutation | CGT,TGT | R,C 484 | XP_006723188.1 | |
| XM_006723126.2 | 1607 | Missense Mutation | CGT,TGT | R,C 430 | XP_006723189.1 | |
| XM_011526694.2 | 1607 | Missense Mutation | CGT,TGT | R,C 453 | XP_011524996.1 | |
| XM_011526695.2 | 1607 | Missense Mutation | CGT,TGT | R,C 440 | XP_011524997.1 | |
| XM_011526698.2 | 1607 | Missense Mutation | CGT,TGT | R,C 262 | XP_011525000.1 | |
| XM_011526700.2 | 1607 | Missense Mutation | CGT,TGT | R,C 168 | XP_011525002.1 | |
| XM_011526701.2 | 1607 | Missense Mutation | CGT,TGT | R,C 99 | XP_011525003.1 | |
| XM_017026550.1 | 1607 | Missense Mutation | CGT,TGT | R,C 252 | XP_016882039.1 | |
| XM_017026551.1 | 1607 | Missense Mutation | CGT,TGT | R,C 485 | XP_016882040.1 | |