Search
Search

CBARP
STK11GCCCGCAAGGCCTGCTCCGCCAGC[AG/CC]CAAGATCCGCCGGCTGTCGGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
CBARP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| CBARP - CACN beta subunit associated regulatory protein | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| STK11 - serine/threonine kinase 11 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000455.4 | 2015 | Missense Mutation | NP_000446.1 | |||
| XM_005259617.2 | 2015 | Silent Mutation | XP_005259674.1 | |||
| XM_005259618.3 | 2015 | Intron | XP_005259675.1 | |||
| XM_011528209.1 | 2015 | Silent Mutation | XP_011526511.1 | |||