Search
Search

GNAZ
RSPH14GCCTCACTGTCTGCCTGGTTTTTGT[C/T]TGTCTGCCCCTTTGTCTCCCAGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GNAZ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GNAZ - G protein subunit alpha z | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_002073.2 | 13698 | Intron | NP_002064.1 | |||
| XM_017028786.1 | 13698 | UTR 3 | XP_016884275.1 | |||
| RSPH14 - radial spoke head 14 homolog | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_014433.2 | 13698 | Intron | NP_055248.1 | |||
| XM_011530149.2 | 13698 | Intron | XP_011528451.1 | |||
| XM_011530150.1 | 13698 | Intron | XP_011528452.1 | |||
| XM_011530151.2 | 13698 | Intron | XP_011528453.1 | |||
| XM_011530152.2 | 13698 | Intron | XP_011528454.1 | |||
| XM_011530154.2 | 13698 | Intron | XP_011528456.1 | |||
| XM_011530155.2 | 13698 | Intron | XP_011528457.1 | |||
| XM_017028774.1 | 13698 | Intron | XP_016884263.1 | |||