Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_353164355_10
          See other FAM3A GT Assays ›
          SNP ID:
          rs375568337
          Gene
          FAM3A
          Gene Name
          family with sequence similarity 3 member A
          Set Membership:
          -
          Chromosome Location:
          Chr.X: 154508485 - 154508485 on Build GRCh38
          Polymorphism:
          G/A, Transition substitution
          Context Sequence [VIC/FAM]:

          CCCAGGGAGCTCACATCTTGTCCTC[G/A]AGGCAGATCTTGGGCCCAATGACGT

          Assay ID C_353164355_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 300492

          Literature Links:

          FAM3A PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          FAM3A - family with sequence similarity 3 member A
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001171132.2 955 Silent Mutation CTC,CTT L,L 88 NP_001164603.1
          NM_001171133.2 955 Silent Mutation CTC,CTT L,L 50 NP_001164604.1
          NM_001171134.2 955 Silent Mutation CTC,CTT L,L 88 NP_001164605.1
          NM_001282311.1 955 Silent Mutation CTC,CTT L,L 102 NP_001269240.1
          NM_001282312.1 955 Silent Mutation CTC,CTT L,L 88 NP_001269241.1
          NM_021806.3 955 Silent Mutation CTC,CTT L,L 88 NP_068578.2
          XM_005274714.3 955 Silent Mutation CTC,CTT L,L 102 XP_005274771.1
          XM_005274716.3 955 Silent Mutation CTC,CTT L,L 102 XP_005274773.1
          XM_005277879.4 955 Silent Mutation CTC,CTT L,L 88 XP_005277936.1
          XM_006724831.2 955 Silent Mutation CTC,CTT L,L 102 XP_006724894.1
          XM_006724832.3 955 Silent Mutation CTC,CTT L,L 88 XP_006724895.1
          XM_006724833.2 955 Silent Mutation CTC,CTT L,L 102 XP_006724896.1
          XM_006724834.2 955 Silent Mutation CTC,CTT L,L 47 XP_006724897.1
          XM_011531185.2 955 Silent Mutation CTC,CTT L,L 47 XP_011529487.1
          XM_017029699.1 955 Silent Mutation CTC,CTT L,L 47 XP_016885188.1
          XM_017029700.1 955 Silent Mutation CTC,CTT L,L 47 XP_016885189.1
          XM_017029701.1 955 Silent Mutation CTC,CTT L,L 47 XP_016885190.1
          XM_017029702.1 955 Silent Mutation CTC,CTT L,L 47 XP_016885191.1

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0