Search
Search

MORC4AAGTAAGTCTCTCCTTATCAGTAAT[T/C]ACTTTAATGTAAATGGATTAAATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MORC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MORC4 - MORC family CW-type zinc finger 4 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001085354.2 | Intron | NP_001078823.1 | ||||
| NM_024657.4 | Intron | NP_078933.3 | ||||
| XM_005262190.3 | Intron | XP_005262247.1 | ||||
| XM_006724691.2 | Intron | XP_006724754.1 | ||||
| XM_011531027.2 | Intron | XP_011529329.1 | ||||
| XM_017029844.1 | Intron | XP_016885333.1 | ||||