Search
Search

IPO9-AS1
MIR1231
NAV1TTGGCCCTAGAACTTCGGATCAAGA[G/T]ACAAAACTCCTCAGATAGCATCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
IPO9-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| IPO9-AS1 - IPO9 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR1231 - microRNA 1231 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| NAV1 - neuron navigator 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001167738.1 | 3388 | Missense Mutation | AGA,ATA | R,I 787 | NP_001161210.1 | |
| NM_020443.4 | 3388 | Missense Mutation | AGA,ATA | R,I 1178 | NP_065176.3 | |
| XM_006711609.2 | 3388 | Missense Mutation | AGA,ATA | R,I 787 | XP_006711672.2 | |
| XM_006711610.2 | 3388 | Missense Mutation | AGA,ATA | R,I 779 | XP_006711673.2 | |
| XM_006711611.1 | 3388 | Missense Mutation | AGA,ATA | R,I 686 | XP_006711674.1 | |
| XM_011510097.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1178 | XP_011508399.1 | |
| XM_011510098.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1170 | XP_011508400.1 | |
| XM_011510099.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1170 | XP_011508401.1 | |
| XM_011510100.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1121 | XP_011508402.1 | |
| XM_011510101.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1113 | XP_011508403.1 | |
| XM_011510102.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1113 | XP_011508404.1 | |
| XM_017002751.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1121 | XP_016858240.1 | |
| XM_017002752.1 | 3388 | Missense Mutation | AGA,ATA | R,I 779 | XP_016858241.1 | |
| XM_017002753.1 | 3388 | Missense Mutation | AGA,ATA | R,I 1170 | XP_016858242.1 | |
| XM_017002754.1 | 3388 | Intron | XP_016858243.1 | |||