Search
Search

ZNF436
ZNF436-AS1AGCTGGAGTTCCTCGGAACGGGAGG[A/G]CTCGCAGCTCTCCCTTTCCCCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ZNF436 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ZNF436 - zinc finger protein 436 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001077195.1 | 152 | UTR 5 | NP_001070663.1 | |||
| NM_030634.2 | 152 | Intron | NP_085137.1 | |||
| XM_011542201.1 | 152 | UTR 5 | XP_011540503.1 | |||
| ZNF436-AS1 - ZNF436 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||