Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_362921285_10
          See other AXDND1 GT Assays ›
          SNP ID:
          rs758610430
          Gene
          AXDND1
          Gene Name
          axonemal dynein light chain domain containing 1
          Set Membership:
          -
          Chromosome Location:
          Chr.1: 179509358 - 179509358 on Build GRCh38
          Polymorphism:
          A/C, Transversion substitution
          Context Sequence [VIC/FAM]:

          TCCTTTCTAATATCAAAGGCAGAAA[A/C]ATTACATTATTGACATATGAAGAAA

          Assay ID C_362921285_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          4 submissions

          Phenotype:

          Literature Links:

          AXDND1 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          AXDND1 - axonemal dynein light chain domain containing 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_144696.5 1785 Missense Mutation AAA,AAC K,N 817 NP_653297.3
          XM_011509165.1 1785 Missense Mutation AAA,AAC K,N 817 XP_011507467.1
          XM_011509166.2 1785 Missense Mutation AAA,AAC K,N 817 XP_011507468.1
          XM_011509167.2 1785 Missense Mutation AAA,AAC K,N 817 XP_011507469.1
          XM_011509168.1 1785 Missense Mutation AAA,AAC K,N 817 XP_011507470.1
          XM_011509169.1 1785 Missense Mutation AAA,AAC K,N 796 XP_011507471.1
          XM_011509170.1 1785 Intron XP_011507472.1
          XM_011509171.1 1785 Missense Mutation AAA,AAC K,N 775 XP_011507473.1
          XM_011509174.1 1785 Intron XP_011507476.1
          XM_011509175.1 1785 Intron XP_011507477.1
          XM_011509176.1 1785 Missense Mutation AAA,AAC K,N 718 XP_011507478.1
          XM_011509178.2 1785 Intron XP_011507480.1
          XM_011509179.2 1785 Missense Mutation AAA,AAC K,N 605 XP_011507481.1
          XM_011509180.1 1785 Intron XP_011507482.1
          XM_011509181.2 1785 Missense Mutation AAA,AAC K,N 458 XP_011507483.1
          XM_017000257.1 1785 Missense Mutation AAA,AAC K,N 570 XP_016855746.1
          XM_017000258.1 1785 Missense Mutation AAA,AAC K,N 524 XP_016855747.1
          XM_017000259.1 1785 Missense Mutation AAA,AAC K,N 302 XP_016855748.1

          Back To Top

          More Information



          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline