Search
Search

MTMR11
OTUD7BAATAAAGCAAACGGAATTGGGATTG[G/A]CTAACTTATAAATAAAAACAAACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MTMR11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MTMR11 - myotubularin related protein 11 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| OTUD7B - OTU deubiquitinase 7B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020205.3 | 5341 | UTR 3 | NP_064590.2 | |||
| XM_011509782.1 | 5341 | UTR 3 | XP_011508084.1 | |||
| XM_011509783.2 | 5341 | UTR 3 | XP_011508085.1 | |||
| XM_011509784.2 | 5341 | UTR 3 | XP_011508086.1 | |||
| XM_011509785.2 | 5341 | UTR 3 | XP_011508087.1 | |||
| XM_011509786.1 | 5341 | UTR 3 | XP_011508088.1 | |||
| XM_011509787.1 | 5341 | UTR 3 | XP_011508089.1 | |||
| XM_011509788.1 | 5341 | UTR 3 | XP_011508090.1 | |||
| XM_017001850.1 | 5341 | UTR 3 | XP_016857339.1 | |||
| XM_017001851.1 | 5341 | UTR 3 | XP_016857340.1 | |||