Search
Search

AHDC1CCCACCGCCCCGTCCACCTCGCGGC[T/-]GCCGGGGCCCAGTGCTCCGTTTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
AHDC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| AHDC1 - AT-hook DNA binding motif containing 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001029882.3 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | NP_001025053.1 | |
| XM_005245848.3 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_005245905.1 | |
| XM_005245849.3 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_005245906.1 | |
| XM_005245850.3 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_005245907.1 | |
| XM_005245851.3 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_005245908.1 | |
| XM_005245852.3 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_005245909.1 | |
| XM_011541255.1 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_011539557.1 | |
| XM_011541256.2 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_011539558.1 | |
| XM_011541257.2 | 4872 | Frame Shift Delete | CAG,CGC | Q,R 1270 | XP_011539559.1 | |