Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_371286185_10
          See other BIRC6 GT Assays ›
          SNP ID:
          rs762532105
          Gene
          BIRC6
          Gene Name
          baculoviral IAP repeat containing 6
          Set Membership:
          -
          Chromosome Location:
          Chr.2: 32370811 - 32370811 on Build GRCh38
          Polymorphism:
          T/C, Transition substitution
          Context Sequence [VIC/FAM]:

          GACTTTGAATGATCTTTGCATATAG[T/C]ACCTTTTAATAGGCTATTTTAAAAT

          Assay ID C_371286185_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 605638

          Literature Links:

          BIRC6 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          BIRC6 - baculoviral IAP repeat containing 6
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_016252.3 Intron NP_057336.3
          XM_005264449.4 Intron XP_005264506.2
          XM_005264450.4 Intron XP_005264507.2
          XM_005264451.4 Intron XP_005264508.2
          XM_005264452.4 Intron XP_005264509.2
          XM_005264453.4 Intron XP_005264510.2
          XM_005264454.4 Intron XP_005264511.2
          XM_005264455.4 Intron XP_005264512.2
          XM_006712054.3 Intron XP_006712117.1
          XM_006712055.3 Intron XP_006712118.1
          XM_006712056.3 Intron XP_006712119.1
          XM_011533003.2 Intron XP_011531305.1
          XM_011533005.2 Intron XP_011531307.1
          XM_017004556.1 Intron XP_016860045.1
          XM_017004557.1 Intron XP_016860046.1
          XM_017004558.1 Intron XP_016860047.1
          XM_017004559.1 Intron XP_016860048.1
          XM_017004560.1 Intron XP_016860049.1

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          apoptotic process
          mitotic nuclear division
          positive regulation of cell proliferation
          negative regulation of endopeptidase activity
          protein ubiquitination
          regulation of cytokinesis
          regulation of cell proliferation
          negative regulation of apoptotic process
          cell division
          labyrinthine layer development
          spongiotrophoblast layer development
          negative regulation of extrinsic apoptotic signaling pathway
          ubiquitin-protein transferase activity
          cysteine-type endopeptidase inhibitor activity
          protein binding
          ligase activity
          ubiquitin conjugating enzyme activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0