Search
Search

HOXD-AS2
HOXD10
HOXD3
HOXD8
HOXD9
LOC100129455TGGAAAAGAAAGGCTTTGATTTGCT[T/C]TGATAACGCCAGGATCTGGGGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
HOXD-AS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| HOXD-AS2 - HOXD cluster antisense RNA 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HOXD10 - homeobox D10 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HOXD3 - homeobox D3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HOXD8 - homeobox D8 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001199746.1 | Intron | NP_001186675.1 | ||||
| NM_001199747.1 | Intron | NP_001186676.1 | ||||
| NM_019558.3 | Intron | NP_062458.1 | ||||
| HOXD9 - homeobox D9 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC100129455 - uncharacterized LOC100129455 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||