Search
Search

LOC101927055
TTNGTTAAAATCACTGAAATTGAAGATC[T/C]GCCTGCCCTGTTTTGGGCAACCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
LOC101927055 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| LOC101927055 - uncharacterized LOC101927055 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| TTN - titin | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001256850.1 | 4840 | Missense Mutation | AGA,GGA | R,G 1538 | NP_001243779.1 | |
| NM_001267550.2 | 4840 | Missense Mutation | AGA,GGA | R,G 1538 | NP_001254479.2 | |
| NM_003319.4 | 4840 | Missense Mutation | AGA,GGA | R,G 1492 | NP_003310.4 | |
| NM_133378.4 | 4840 | Missense Mutation | AGA,GGA | R,G 1538 | NP_596869.4 | |
| NM_133379.4 | 4840 | Missense Mutation | AGA,GGA | R,G 1538 | NP_596870.2 | |
| NM_133432.3 | 4840 | Missense Mutation | AGA,GGA | R,G 1492 | NP_597676.3 | |
| NM_133437.4 | 4840 | Missense Mutation | AGA,GGA | R,G 1492 | NP_597681.4 | |
| XM_017004819.1 | 4840 | Missense Mutation | AGA,GGA | R,G 1539 | XP_016860308.1 | |
| XM_017004820.1 | 4840 | Missense Mutation | AGA,GGA | R,G 1539 | XP_016860309.1 | |
| XM_017004821.1 | 4840 | Missense Mutation | AGA,GGA | R,G 1538 | XP_016860310.1 | |
| XM_017004822.1 | 4840 | Missense Mutation | AGA,GGA | R,G 1539 | XP_016860311.1 | |
| XM_017004823.1 | 4840 | Missense Mutation | AGA,GGA | R,G 1539 | XP_016860312.1 | |