Search
Search

LOC100421494
SOWAHBCCTTTGCACTAGCAATTTGATGACC[C/G]CCTGGTGGCCGTGAATGGCTGCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC100421494 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| LOC100421494 - microtubule affinity regulating kinase 2 pseudogene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SOWAHB - sosondowah ankyrin repeat domain family member B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001029870.1 | 2051 | Missense Mutation | GCG,GGG | A,G 684 | NP_001025041.1 | |