Search
Search

MYOTTCAGTTAGACCTAGGACTCTCTATG[G/A]TGTCTACACGGACAGGGAATATATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MYOT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MYOT - myotilin | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001135940.1 | Intron | NP_001129412.1 | ||||
| NM_001300911.1 | Intron | NP_001287840.1 | ||||
| NM_006790.2 | Intron | NP_006781.1 | ||||
| XM_017010060.1 | Intron | XP_016865549.1 | ||||
| XM_017010061.1 | Intron | XP_016865550.1 | ||||
| XM_017010062.1 | Intron | XP_016865551.1 | ||||