Search
Search

HIST1H2AH
HIST1H2BK
HIST1H4I
MIR3143GGAAAACAAAGGCTCTTTTCAGAGC[C/T]ATTCTACACTGTCTTAGAGAAAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
HIST1H2AH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| HIST1H2AH - histone cluster 1, H2ah | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_080596.2 | Intron | NP_542163.1 | ||||
| HIST1H2BK - histone cluster 1, H2bk | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001312653.1 | Intron | NP_001299582.1 | ||||
| NM_080593.2 | Intron | NP_542160.1 | ||||
| HIST1H4I - histone cluster 1, H4i | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR3143 - microRNA 3143 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||