Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_403981876_10
          See other IMMP2L GT Assays ›
          SNP ID:
          rs769972573
          Gene
          IMMP2L LRRN3
          Gene Name
          inner mitochondrial membrane peptidase subunit 2
          leucine rich repeat neuronal 3
          Set Membership:
          -
          Chromosome Location:
          Chr.7: 111104602 - 111104602 on Build GRCh38
          Polymorphism:
          T/C, Transition substitution
          Context Sequence [VIC/FAM]:

          CATGCTAAATAACAGAAAAGTCTCA[T/C]CATTAATCCCCCTATCTAGTCTTTT

          Assay ID C_403981876_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 605977

          Literature Links:

          IMMP2L PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          IMMP2L - inner mitochondrial membrane peptidase subunit 2
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001244606.1 Intron NP_001231535.1
          NM_032549.3 Intron NP_115938.1
          XM_005250630.3 Intron XP_005250687.1
          XM_011516605.2 Intron XP_011514907.1
          XM_011516608.2 Intron XP_011514910.1
          XM_011516609.2 Intron XP_011514911.1
          XM_011516611.2 Intron XP_011514913.1
          XM_011516613.2 Intron XP_011514915.1
          XM_017012699.1 Intron XP_016868188.1
          XM_017012700.1 Intron XP_016868189.1
          XM_017012701.1 Intron XP_016868190.1
          XM_017012702.1 Intron XP_016868191.1
          XM_017012703.1 Intron XP_016868192.1
          XM_017012704.1 Intron XP_016868193.1
          XM_017012705.1 Intron XP_016868194.1
          LRRN3 - leucine rich repeat neuronal 3
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001099658.1 Intron NP_001093128.1
          NM_001099660.1 Intron NP_001093130.1
          NM_018334.4 Intron NP_060804.3

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          signal peptide processing
          protein processing involved in protein targeting to mitochondrion
          superoxide metabolic process
          cellular response to DNA damage stimulus
          spermatogenesis
          brain development
          blood circulation
          respiratory electron transport chain
          ovulation
          mitochondrial respiratory chain complex assembly
          cerebellum vasculature development
          serine-type endopeptidase activity
          peptidase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline