Search
Search

PHTF2
TMEM60CAAAGATAATTTTGCTTCATGAGCC[C/T]ACATATTTATTCTTCTTCATAATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
PHTF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| PHTF2 - putative homeodomain transcription factor 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001127357.1 | Intron | NP_001120829.1 | ||||
| NM_001127358.1 | Intron | NP_001120830.1 | ||||
| NM_001127359.1 | Intron | NP_001120831.1 | ||||
| NM_001127360.1 | Intron | NP_001120832.1 | ||||
| NM_020432.4 | Intron | NP_065165.3 | ||||
| XM_005250508.4 | Intron | XP_005250565.1 | ||||
| XM_005250509.4 | Intron | XP_005250566.1 | ||||
| XM_006716063.3 | Intron | XP_006716126.1 | ||||
| XM_011516422.2 | Intron | XP_011514724.1 | ||||
| XM_011516423.2 | Intron | XP_011514725.1 | ||||
| XM_011516424.2 | Intron | XP_011514726.1 | ||||
| XM_017012458.1 | Intron | XP_016867947.1 | ||||
| XM_017012459.1 | Intron | XP_016867948.1 | ||||
| TMEM60 - transmembrane protein 60 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_032936.3 | Intron | NP_116325.1 | ||||