Search
Search

CYP2D6
CYP2D7
LOC102723722
NDUFA6-AS1CCGGGCAGTGGCAGGGGGCCTGGTG[A/G]GTAGCGTGCAGCCCAGCGTTGGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2D6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| CYP2D6 - cytochrome P450 family 2 subfamily D member 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000106.5 | 190 | Missense Mutation | CCA,TCA | P,S 34 | NP_000097.3 | |
| NM_001025161.2 | 190 | Missense Mutation | CCA,TCA | P,S 34 | NP_001020332.2 | |
| XM_011529966.2 | 190 | Intron | XP_011528268.1 | |||
| XM_011529968.2 | 190 | Intron | XP_011528270.1 | |||
| XM_011529970.2 | 190 | Intron | XP_011528272.1 | |||
| XM_011529972.2 | 190 | Intron | XP_011528274.1 | |||
| CYP2D7 - cytochrome P450 family 2 subfamily D member 7 (gene/pseudogene) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC102723722 - uncharacterized LOC102723722 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| XM_017029174.1 | 190 | Intron | XP_016884663.1 | |||
| NDUFA6-AS1 - NDUFA6 antisense RNA 1 (head to head) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
Set Membership: |
|