Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__11668955_10
          See other LIPC GT Assays ›
          SNP ID:
          rs1869145
          Gene
          LIPC LIPC-AS1
          Gene Name
          lipase C, hepatic type
          LIPC antisense RNA 1
          Set Membership:
          > HapMap
          Chromosome Location:
          Chr.15: 58505300 - 58505300 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          CCAGCCCCCCTACTCACCAGCCATG[C/T]GGCCTCAGGCAAGTGGGATAATCCC

          Assay ID C__11668955_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 151670

          Literature Links:

          LIPC PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          C (0.27)
          (0.73)
          Caucasian - Not Available CEPH (CEU)
          C (0.21)
          (0.79)
          EAS
          C (0.24)
          (0.76)
          African American - Not Available YRI (Yoruba)
          C (0.19)
          (0.81)
          SAS
          C (0.41)
          (0.59)
          Chinese - Not Available JPT (Japanese)
          C (0.27)
          (0.73)
          AFR
          C (0.20)
          (0.80)
          Japanese - Not Available CHB (Han Chinese)
          C (0.23)
          (0.77)
          EUR
          C (0.25)
          (0.75)
          AMR
          C (0.26)
          (0.74)
          LIPC - lipase C, hepatic type
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_000236.2 Intron NP_000227.2
          XM_005254372.1 Intron XP_005254429.1
          XM_005254374.4 Intron XP_005254431.2
          XM_006720502.3 Intron XP_006720565.1
          XM_017022176.1 Intron XP_016877665.1
          LIPC-AS1 - LIPC antisense RNA 1
          There are no transcripts associated with this gene.

          Back To Top

          More Information


          Set Membership:

          HapMap

          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          liver development
          fatty acid biosynthetic process
          phosphatidylserine metabolic process
          circadian rhythm
          cholesterol metabolic process
          response to carbohydrate
          response to acetate
          heparan sulfate proteoglycan biosynthetic process
          triglyceride catabolic process
          response to nutrient levels
          response to magnesium ion
          cellular response to hormone stimulus
          chylomicron remodeling
          very-low-density lipoprotein particle remodeling
          intermediate-density lipoprotein particle remodeling
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          chylomicron remnant clearance
          low-density lipoprotein particle clearance
          phosphatidylcholine catabolic process
          response to drug
          cholesterol homeostasis
          response to amino acid
          response to peptide hormone
          reverse cholesterol transport
          lipid digestion
          phosphatidylethanolamine metabolic process
          phosphatidic acid metabolic process
          glycerophospholipid catabolic process
          response to copper ion
          developmental growth
          protein oligomerization
          response to glucocorticoid
          response to calcium ion
          triglyceride homeostasis
          response to fatty acid
          phospholipase activity
          lysophospholipase activity
          triglyceride lipase activity
          heparin binding
          lipid binding
          phosphatidylcholine 1-acylhydrolase activity
          transferase activity, transferring acyl groups
          low-density lipoprotein particle binding
          apolipoprotein binding
          heparan sulfate proteoglycan binding
          acylglycerol lipase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline