Search
Search

RUVBL1
RUVBL1-AS1
SEC61A1GAGATGGAAGCATGTGGCCAGGCGG[A/G]TAGGAGACAGTGCAGTAAGGGTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUVBL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| RUVBL1 - RuvB like AAA ATPase 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001319084.1 | Intron | NP_001306013.1 | ||||
| NM_001319086.1 | Intron | NP_001306015.1 | ||||
| NM_003707.2 | Intron | NP_003698.1 | ||||
| XM_005247841.3 | Intron | XP_005247898.1 | ||||
| XM_011513248.1 | Intron | XP_011511550.1 | ||||
| XM_011513249.2 | Intron | XP_011511551.1 | ||||
| XM_017007356.1 | Intron | XP_016862845.1 | ||||
| XM_017007357.1 | Intron | XP_016862846.1 | ||||
| RUVBL1-AS1 - RUVBL1 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SEC61A1 - Sec61 translocon alpha 1 subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_013336.3 | Intron | NP_037468.1 | ||||
Set Membership: |
|