Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600946 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GHR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GHR - growth hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000163.4 | Intron | NP_000154.1 | ||||
NM_001242399.2 | Intron | NP_001229328.1 | ||||
NM_001242400.2 | Intron | NP_001229329.1 | ||||
NM_001242401.3 | Intron | NP_001229330.1 | ||||
NM_001242402.2 | Intron | NP_001229331.1 | ||||
NM_001242403.2 | Intron | NP_001229332.1 | ||||
NM_001242404.2 | Intron | NP_001229333.1 | ||||
NM_001242405.2 | Intron | NP_001229334.1 | ||||
NM_001242406.2 | Intron | NP_001229335.1 | ||||
NM_001242460.1 | Intron | NP_001229389.1 | ||||
NM_001242462.1 | Intron | NP_001229391.1 |
Set Membership: |
HapMap |