Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607585 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATM - ATM serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000051.3 | 1364 | Intron | NP_000042.3 | |||
XM_005271561.4 | 1364 | Intron | XP_005271618.2 | |||
XM_005271562.4 | 1364 | Intron | XP_005271619.2 | |||
XM_006718843.3 | 1364 | Intron | XP_006718906.1 | |||
XM_006718845.1 | 1364 | Intron | XP_006718908.1 | |||
XM_011542840.2 | 1364 | Intron | XP_011541142.1 | |||
XM_011542842.2 | 1364 | Intron | XP_011541144.1 | |||
XM_011542843.2 | 1364 | Intron | XP_011541145.1 | |||
XM_011542844.2 | 1364 | Intron | XP_011541146.1 | |||
XM_011542845.1 | 1364 | Intron | XP_011541147.1 | |||
XM_017017789.1 | 1364 | Intron | XP_016873278.1 | |||
XM_017017790.1 | 1364 | Intron | XP_016873279.1 | |||
XM_017017791.1 | 1364 | Intron | XP_016873280.1 | |||
XM_017017792.1 | 1364 | Intron | XP_016873281.1 |
C11orf65 - chromosome 11 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152587.3 | 1364 | Intron | NP_689800.3 | |||
XM_005271412.3 | 1364 | Intron | XP_005271469.1 | |||
XM_005271413.3 | 1364 | Intron | XP_005271470.1 | |||
XM_005271414.3 | 1364 | Intron | XP_005271471.1 | |||
XM_005271415.4 | 1364 | Intron | XP_005271472.1 | |||
XM_011542639.2 | 1364 | Intron | XP_011540941.1 | |||
XM_011542640.2 | 1364 | Intron | XP_011540942.1 | |||
XM_011542641.2 | 1364 | Intron | XP_011540943.1 | |||
XM_011542642.1 | 1364 | Intron | XP_011540944.1 | |||
XM_011542643.2 | 1364 | Intron | XP_011540945.1 | |||
XM_017017246.1 | 1364 | Intron | XP_016872735.1 | |||
XM_017017247.1 | 1364 | Intron | XP_016872736.1 | |||
XM_017017248.1 | 1364 | Intron | XP_016872737.1 | |||
XM_017017249.1 | 1364 | UTR 3 | XP_016872738.1 | |||
XM_017017250.1 | 1364 | UTR 3 | XP_016872739.1 |
Set Membership: |
HapMap |