Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615931 | ||||||||||||||||||||
Literature Links: |
PRR16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRR16 - proline rich 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300783.1 | Intron | NP_001287712.1 | ||||
NM_001308087.1 | Intron | NP_001295016.1 | ||||
NM_001308088.1 | Intron | NP_001295017.1 | ||||
NM_016644.2 | Intron | NP_057728.1 | ||||
XM_011543452.2 | Intron | XP_011541754.1 | ||||
XM_011543453.2 | Intron | XP_011541755.1 | ||||
XM_011543454.2 | Intron | XP_011541756.1 |